Assay Details
Gene Symbol: LIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214402.2 | NP_999567.1 | 2 - 3 | 204 | 104 | |
XM_005670785.3 | XP_005670842.1 | 1639 | 104 | |||
XM_013983025.2 | XP_013838479.1 | 2329 | 104 | |||
XM_013983026.2 | XP_013838480.1 | 2056 | 104 | |||
GenBank mRNA | AY585336.1 | - | 2 - 3 | 204 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
