Assay Details
Gene Symbol: CCL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024589.1 | NP_001019760.1 | 1 - 2 | 114 | 60 | |
GenBank mRNA | AJ577084.1 | - | 1 - 2 | 114 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
