Assay Details
Gene Symbol: FABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025229.1 | NP_001020400.1 | 3 - 4 | 433 | 117 | |
GenBank mRNA | AK391779.1 | - | 3 - 4 | 451 | 117 | |
AY610347.1 | - | 3 - 4 | 433 | 117 | ||
DQ000160.1 | - | 3 - 4 | 433 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
