Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030535.1 | NP_001025706.1 | 1 - 2 | 219 | 92 | |
XM_005664129.3 | XP_005664186.1 | 564 | 92 | |||
XM_005664130.3 | XP_005664187.1 | 593 | 92 | |||
XM_021091203.1 | XP_020946862.1 | 579 | 92 | |||
GenBank mRNA | AK234400.1 | - | 3 - 4 | 373 | 92 | |
AY521564.1 | - | 3 - 4 | 357 | 92 | ||
DQ087226.1 | - | 1 - 2 | 219 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
