Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030535.1 | NP_001025706.1 | 2 - 3 | 352 | 111 | |
XM_005664129.3 | XP_005664186.1 | 697 | 111 | |||
XM_005664130.3 | XP_005664187.1 | 726 | 111 | |||
XM_021091203.1 | XP_020946862.1 | 712 | 111 | |||
GenBank mRNA | AK234400.1 | - | 4 - 5 | 506 | 111 | |
AK396188.1 | - | 4 - 5 | 512 | 111 | ||
AY521564.1 | - | 4 - 5 | 490 | 111 | ||
DQ087226.1 | - | 2 - 3 | 352 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
