Assay Details
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037961.1 | NP_001033050.1 | 4 - 5 | 573 | 58 | |
GenBank mRNA | AK231242.1 | - | 4 - 5 | 419 | 58 | |
AK392820.1 | - | 4 - 5 | 642 | 58 | ||
DQ295031.1 | - | 4 - 5 | 573 | 58 | ||
DQ333333.1 | - | 4 - 5 | 431 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
