Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213861.1 | NP_999026.1 | 2 - 3 | 204 | 99 | |
XM_021100436.1 | XP_020956095.1 | 237 | 99 | |||
GenBank mRNA | AB194099.1 | - | 2 - 3 | 144 | 99 | |
AY294018.1 | - | 2 - 3 | 226 | 99 | ||
EU139160.1 | - | 2 - 3 | 227 | 99 | ||
EU249805.1 | - | 2 - 3 | 204 | 99 | ||
FJ543109.1 | - | 2 - 3 | 204 | 99 | ||
JF906511.1 | - | 2 - 3 | 204 | 99 | ||
X56750.1 | - | 204 | 99 | |||
X58428.1 | - | 231 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
