Assay Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213869.1 | NP_999034.2 | 4 - 5 | 574 | 150 | |
GenBank mRNA | AK349245.1 | - | 4 - 5 | 508 | 150 | |
AY536527.1 | - | 3 - 4 | 454 | 150 | ||
DQ415717.1 | - | 3 - 4 | 447 | 150 | ||
EU390781.1 | - | 3 - 4 | 447 | 150 | ||
EU400158.1 | - | 3 - 4 | 447 | 150 | ||
HM362439.1 | - | 3 - 4 | 456 | 150 | ||
JF276447.1 | - | 3 - 4 | 447 | 150 | ||
JF742642.1 | - | 3 - 4 | 447 | 150 | ||
JQ177096.1 | - | 3 - 4 | 456 | 150 | ||
M22761.1 | - | 4 - 5 | 571 | 150 | ||
M27326.1 | - | 3 - 4 | 150 | |||
U19787.1 | - | 4 - 5 | 574 | 150 | ||
U19788.1 | - | 4 - 5 | 493 | 150 | ||
X53325.1 | - | 456 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
