Assay Details
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214123.1 | NP_999288.1 | 1 - 2 | 163 | 123 | |
GenBank mRNA | HQ236500.1 | - | 1 - 2 | 140 | 123 | |
JF906512.1 | - | 1 - 2 | 140 | 123 | ||
X68330.1 | - | 163 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
