Assay Details
Gene Symbol: MBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214125.1 | NP_999290.1 | 1 - 2 | 180 | 82 | |
GenBank mRNA | AF164576.1 | - | 1 - 2 | 180 | 82 | |
KF803270.1 | - | 1 - 2 | 180 | 82 | ||
KF803271.1 | - | 1 - 2 | 180 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
