Assay Details
Gene Symbol: PRDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214144.1 | NP_999309.1 | 5 - 6 | 553 | 64 | |
GenBank mRNA | AF110735.2 | - | 5 - 6 | 553 | 64 | |
AK231569.1 | - | 5 - 6 | 543 | 64 | ||
AK233717.1 | - | 5 - 6 | 546 | 64 | ||
AK234018.1 | - | 5 - 6 | 478 | 64 | ||
AK392392.1 | - | 5 - 6 | 596 | 64 | ||
AK399969.1 | - | 5 - 6 | 609 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
