Assay Details
Gene Symbol: AMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214195.1 | NP_999360.1 | 7 - 8 | 1099 | 63 | |
XM_005663621.3 | XP_005663678.2 | 1322 | 63 | |||
XM_005663623.3 | XP_005663680.2 | 1408 | 63 | |||
XM_021088708.1 | XP_020944367.1 | 1404 | 63 | |||
XM_021088709.1 | XP_020944368.1 | 1528 | 63 | |||
XM_021088710.1 | XP_020944369.1 | 1221 | 63 | |||
XM_021088711.1 | XP_020944370.1 | 1164 | 63 | |||
GenBank mRNA | AF064742.1 | - | 7 - 8 | 1099 | 63 | |
AK395394.1 | - | 8 - 9 | 1260 | 63 |
Gene Symbol: LOC100153854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005663628.3 | XP_005663685.2 | 1205 | 63 | ||
GenBank mRNA | AK395638.1 | - | 8 - 9 | 1250 | 63 |
Gene Symbol: LOC100521789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125881.6 | XP_003125929.2 | 1450 | 63 | ||
XM_005663625.3 | XP_005663682.2 | 1282 | 63 | |||
XM_021090122.1 | XP_020945781.1 | 1388 | 63 | |||
XM_021090123.1 | XP_020945782.1 | 1438 | 63 | |||
XM_021090124.1 | XP_020945783.1 | 1164 | 63 |
Gene Symbol: LOC110258046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081016.1 | XP_020936675.1 | 1303 | 63 | ||
XM_021081017.1 | XP_020936676.1 | 1403 | 63 | |||
XM_021081018.1 | XP_020936677.1 | 1285 | 63 | |||
XM_021081019.1 | XP_020936678.1 | 1300 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
