Assay Details
Gene Symbol: AMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214195.1 | NP_999360.1 | 8 - 9 | 1215 | 65 | |
XM_005663621.3 | XP_005663678.2 | 1438 | 65 | |||
XM_005663623.3 | XP_005663680.2 | 1524 | 65 | |||
XM_021088708.1 | XP_020944367.1 | 1520 | 65 | |||
XM_021088709.1 | XP_020944368.1 | 1644 | 65 | |||
XM_021088710.1 | XP_020944369.1 | 1337 | 65 | |||
XM_021088711.1 | XP_020944370.1 | 1280 | 65 | |||
GenBank mRNA | AF064742.1 | - | 8 - 9 | 1215 | 65 | |
AK395394.1 | - | 9 - 10 | 1376 | 65 |
Gene Symbol: LOC100153854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005663628.3 | XP_005663685.2 | 1321 | 65 | ||
GenBank mRNA | AK230675.1 | - | 9 - 10 | 1768 | 65 | |
AK395638.1 | - | 9 - 10 | 1366 | 65 |
Gene Symbol: LOC100521789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125881.6 | XP_003125929.2 | 1566 | 65 | ||
XM_005663625.3 | XP_005663682.2 | 1398 | 65 | |||
XM_021090122.1 | XP_020945781.1 | 1504 | 65 | |||
XM_021090123.1 | XP_020945782.1 | 1554 | 65 | |||
XM_021090124.1 | XP_020945783.1 | 1280 | 65 |
Gene Symbol: LOC110258046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081016.1 | XP_020936675.1 | 1419 | 65 | ||
XM_021081017.1 | XP_020936676.1 | 1519 | 65 | |||
XM_021081018.1 | XP_020936677.1 | 1401 | 65 | |||
XM_021081019.1 | XP_020936678.1 | 1416 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
