Assay Details
Gene Symbol: AMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214195.1 | NP_999360.1 | 9 - 10 | 1336 | 82 | |
XM_005663621.3 | XP_005663678.2 | 1559 | 82 | |||
XM_005663623.3 | XP_005663680.2 | 1645 | 82 | |||
XM_021088708.1 | XP_020944367.1 | 1641 | 82 | |||
XM_021088709.1 | XP_020944368.1 | 1765 | 82 | |||
XM_021088710.1 | XP_020944369.1 | 1458 | 82 | |||
XM_021088711.1 | XP_020944370.1 | 1401 | 82 | |||
GenBank mRNA | AF064742.1 | - | 9 - 10 | 1336 | 82 | |
AK395394.1 | - | 10 - 11 | 1497 | 82 |
Gene Symbol: LOC100153854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005663628.3 | XP_005663685.2 | 1442 | 82 | ||
GenBank mRNA | AK230675.1 | - | 10 - 11 | 1889 | 82 | |
AK395638.1 | - | 10 - 11 | 1487 | 82 |
Gene Symbol: LOC100521789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125881.6 | XP_003125929.2 | 1687 | 82 | ||
XM_005663625.3 | XP_005663682.2 | 1519 | 82 | |||
XM_021090122.1 | XP_020945781.1 | 1625 | 82 | |||
XM_021090123.1 | XP_020945782.1 | 1675 | 82 | |||
XM_021090124.1 | XP_020945783.1 | 1401 | 82 |
Gene Symbol: LOC110258046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081016.1 | XP_020936675.1 | 1540 | 82 | ||
XM_021081017.1 | XP_020936676.1 | 1640 | 82 | |||
XM_021081018.1 | XP_020936677.1 | 1522 | 82 | |||
XM_021081019.1 | XP_020936678.1 | 1537 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
