Assay Details
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214206.1 | NP_999371.1 | 1 - 2 | 110 | 70 | |
GenBank mRNA | AK232208.1 | - | 1 - 2 | 117 | 70 | |
AK233316.1 | - | 1 - 2 | 126 | 70 | ||
AK391018.1 | - | 1 - 2 | 137 | 70 | ||
AK400768.1 | - | 1 - 2 | 137 | 70 | ||
AY609777.1 | - | 1 - 2 | 208 | 70 | ||
U62253.1 | - | 1 - 2 | 110 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
