Assay Details
Gene Symbol: OPALIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214210.1 | NP_999375.1 | 3 - 4 | 660 | 68 | |
XM_013983587.2 | XP_013839041.1 | 793 | 68 | |||
XM_013983589.2 | XP_013839043.1 | 1043 | 68 | |||
XM_013983590.2 | XP_013839044.1 | 511 | 68 | |||
XM_021072121.1 | XP_020927780.1 | 555 | 68 | |||
GenBank mRNA | AK392173.1 | - | 3 - 4 | 398 | 68 | |
AK400134.1 | - | 3 - 4 | 460 | 68 | ||
AK400143.1 | - | 3 - 4 | 470 | 68 | ||
AK400203.1 | - | 3 - 4 | 324 | 68 | ||
X89480.1 | - | 660 | 68 | |||
Y11026.1 | - | 380 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the Y11026.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
