Assay Details
Gene Symbol: OPALIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214210.1 | NP_999375.1 | 4 - 5 | 782 | 88 | |
XM_013983587.2 | XP_013839041.1 | 915 | 88 | |||
XM_013983589.2 | XP_013839043.1 | 1165 | 88 | |||
XM_013983590.2 | XP_013839044.1 | 633 | 88 | |||
XM_021072121.1 | XP_020927780.1 | 677 | 88 | |||
GenBank mRNA | AK392173.1 | - | 4 - 5 | 520 | 88 | |
AK400134.1 | - | 4 - 5 | 582 | 88 | ||
AK400143.1 | - | 4 - 5 | 592 | 88 | ||
AK400203.1 | - | 4 - 5 | 446 | 88 | ||
X89480.1 | - | 782 | 88 | |||
Y11026.1 | - | 502 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the Y11026.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
