Assay Details
Gene Symbol: RHO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214221.1 | NP_999386.1 | 3 - 4 | 695 | 71 | |
GenBank mRNA | AF008947.1 | - | 3 - 4 | 695 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
