Assay Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214363.1 | NP_999528.1 | 5 - 6 | 408 | 62 | |
GenBank mRNA | AK234793.1 | - | 1603 | 62 | ||
AK236805.1 | - | 5 - 6 | 420 | 62 | ||
AK392759.1 | - | 5 - 6 | 419 | 62 | ||
AY610305.1 | - | 5 - 6 | 391 | 62 | ||
AY610355.1 | - | 5 - 6 | 406 | 62 | ||
X79417.1 | - | 408 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
