Assay Details
Gene Symbol: HYAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214440.1 | NP_999605.1 | 1 - 2 | 934 | 78 | |
XM_005669574.3 | XP_005669631.1 | 1124 | 78 | |||
XM_005669575.3 | XP_005669632.1 | 2208 | 78 | |||
XM_005669576.3 | XP_005669633.1 | 2671 | 78 | |||
XM_013981684.1 | XP_013837138.1 | 1430 | 78 | |||
XM_021068571.1 | XP_020924230.1 | 1043 | 78 | |||
GenBank mRNA | AK343526.1 | - | 2 - 3 | 1093 | 78 | |
AY497544.1 | - | 1 - 2 | 934 | 78 | ||
EU925135.1 | - | 1 - 2 | 903 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
