Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005154.1 | NP_001005154.2 | 2 | 1055 | 147 | |
GenBank mRNA | AY609964.1 | - | 2 | 638 | 147 | |
FJ882404.1 | - | 2 | 599 | 147 | ||
HF549032.1 | - | 2 | 467 | 147 | ||
HF549033.1 | - | 1 | 425 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
