Assay Details
Gene Symbol: ADRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128436.1 | NP_001121908.1 | 1 | 947 | 113 | |
XM_021081158.1 | XP_020936817.1 | 1839 | 113 | |||
GenBank mRNA | AK237368.1 | - | 1 | 1192 | 113 | |
KF023571.1 | - | 1 | 947 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
