Assay Details
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142829.1 | NP_001136301.1 | 3 | 641 | 87 | |
XM_005672558.3 | XP_005672615.2 | 952 | 87 | |||
XM_005672559.3 | XP_005672616.2 | 732 | 87 | |||
XM_013984879.2 | XP_013840333.2 | 845 | 87 | |||
XM_021076524.1 | XP_020932183.1 | 750 | 87 | |||
GenBank mRNA | AY609840.1 | - | 4 | 848 | 87 | |
FJ428538.1 | - | 3 | 641 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
