Assay Details
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143723.1 | NP_001137195.1 | 4 - 5 | 569 | 75 | |
XM_005661427.3 | XP_005661484.2 | 2373 | 75 | |||
XM_005661428.3 | XP_005661485.1 | 612 | 75 | |||
GenBank mRNA | FJ436381.1 | - | 4 - 5 | 569 | 75 | |
FJ436383.1 | - | 3 - 4 | 428 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
