Assay Details
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144842.2 | NP_001138314.1 | 2 - 3 | 212 | 67 | |
GenBank mRNA | AK391822.1 | - | 2 - 3 | 240 | 67 | |
FJ487633.1 | - | 2 - 3 | 213 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
