Assay Details
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170828.1 | NP_001164299.1 | 2 - 3 | 405 | 68 | |
XM_005656037.3 | XP_005656094.3 | 520 | 68 | |||
XM_013988775.2 | XP_013844229.2 | 523 | 68 | |||
GenBank mRNA | GU121641.1 | - | 2 - 3 | 405 | 68 | |
GU373697.1 | - | 2 - 3 | 310 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
