Assay Details
Gene Symbol: LTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185138.1 | NP_001172067.1 | 2 - 3 | 211 | 91 | |
XM_005665780.3 | XP_005665837.1 | 495 | 91 | |||
GenBank mRNA | AB154366.1 | - | 2 - 3 | 211 | 91 | |
AB294210.1 | - | 2 - 3 | 211 | 91 | ||
AK233944.1 | - | 2 - 3 | 214 | 91 | ||
AK234043.1 | - | 2 - 3 | 213 | 91 | ||
AK345069.1 | - | 2 - 3 | 214 | 91 | ||
AK345209.1 | - | 2 - 3 | 213 | 91 | ||
AK346577.1 | - | 2 - 3 | 213 | 91 | ||
AK346655.1 | - | 2 - 3 | 213 | 91 | ||
AK346802.1 | - | 2 - 3 | 213 | 91 | ||
AK346819.1 | - | 2 - 3 | 215 | 91 | ||
AK346962.1 | - | 2 - 3 | 213 | 91 | ||
AK346972.1 | - | 2 - 3 | 213 | 91 | ||
AK347038.1 | - | 2 - 3 | 216 | 91 | ||
AK347059.1 | - | 2 - 3 | 218 | 91 | ||
EU282350.1 | - | 2 - 3 | 215 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
