Assay Details
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185154.1 | NP_001172083.1 | 1 - 2 | 92 | 68 | |
NM_001185155.1 | NP_001172084.1 | 1 - 2 | 93 | 68 | ||
XM_021098125.1 | XP_020953784.1 | 138 | 68 | |||
GenBank mRNA | AK236172.1 | - | 1 - 2 | 95 | 68 | |
AK238937.1 | - | 1 - 2 | 93 | 68 | ||
AK396148.1 | - | 1 - 2 | 91 | 68 | ||
AK398319.1 | - | 1 - 2 | 93 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001185154.1, NM_001185155.1, XM_021098125.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
