Assay Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185184.1 | NP_001172113.1 | 2 - 3 | 266 | 81 | |
XM_021088597.1 | XP_020944256.1 | 280 | 81 | |||
GenBank mRNA | AK231087.1 | - | 2 - 3 | 266 | 81 | |
AK391737.1 | - | 2 - 3 | 268 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
