Assay Details
Gene Symbol: APOA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001186294.1 | NP_001173223.1 | 2 - 3 | 110 | 83 | |
GenBank mRNA | AK233612.1 | - | 2 - 3 | 110 | 83 | |
AK345816.1 | - | 2 - 3 | 108 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
