Assay Details
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190174.1 | NP_001177103.1 | 3 - 4 | 421 | 83 | |
XM_021100259.1 | XP_020955918.1 | 820 | 83 | |||
XM_021100314.1 | XP_020955973.1 | 608 | 83 | |||
XM_021100355.1 | XP_020956014.1 | 1094 | 83 | |||
GenBank mRNA | AK234741.1 | - | 5 - 6 | 606 | 83 | |
AK395077.1 | - | 5 - 6 | 609 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
