Assay Details
Gene Symbol: CDC42SE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190193.1 | NP_001177122.1 | 4 - 5 | 582 | 82 | |
XM_005663461.3 | XP_005663518.1 | 663 | 82 | |||
XM_013997164.2 | XP_013852618.1 | 816 | 82 | |||
XM_021088571.1 | XP_020944230.1 | 513 | 82 | |||
GenBank mRNA | AK239716.1 | - | 4 - 5 | 577 | 82 | |
BW960573.1 | - | 4 - 5 | 582 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
