Assay Details
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190219.1 | NP_001177148.1 | 3 - 4 | 302 | 79 | |
GenBank mRNA | AK232476.1 | - | 3 - 4 | 505 | 79 | |
AK392722.1 | - | 3 - 4 | 307 | 79 | ||
AY609640.1 | - | 3 - 4 | 255 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
