Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190222.1 | NP_001177151.1 | 1 - 2 | 260 | 92 | |
NM_001190224.1 | NP_001177153.1 | 2 - 3 | 336 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190222.1, NM_001190224.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
