Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190222.1 | NP_001177151.1 | 3 - 4 | 528 | 82 | |
NM_001190223.1 | NP_001177152.1 | 3 - 4 | 494 | 82 | ||
NM_001190224.1 | NP_001177153.1 | 4 - 5 | 604 | 82 | ||
GenBank mRNA | AK232872.1 | - | 3 - 4 | 458 | 82 | |
AK233265.1 | - | 4 - 5 | 604 | 82 | ||
AK233963.1 | - | 3 - 4 | 371 | 82 | ||
AK236795.1 | - | 3 - 4 | 513 | 82 | ||
AK238663.1 | - | 3 - 4 | 365 | 82 | ||
AK346585.1 | - | 3 - 4 | 294 | 82 | ||
AK389403.1 | - | 3 - 4 | 528 | 82 | ||
AK394516.1 | - | 3 - 4 | 354 | 82 | ||
AK397522.1 | - | 3 - 4 | 494 | 82 | ||
AK398631.1 | - | 3 - 4 | 354 | 82 | ||
AK398690.1 | - | 3 - 4 | 478 | 82 | ||
AK398918.1 | - | 3 - 4 | 389 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
