Assay Details
Gene Symbol: GPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190240.1 | NP_001177169.1 | 5 - 6 | 865 | 85 | |
XM_021090918.1 | XP_020946577.1 | 930 | 85 | |||
GenBank mRNA | AK232843.1 | - | 5 - 6 | 865 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
