Assay Details
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195368.1 | NP_001182297.1 | 4 - 5 | 708 | 109 | |
XM_013996752.2 | XP_013852206.1 | 987 | 109 | |||
GenBank mRNA | AK344212.1 | - | 5 - 6 | 808 | 109 | |
GU373688.1 | - | 4 - 5 | 708 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
