Assay Details
Gene Symbol: SH3BGRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095627.1 | XP_020951286.1 | 325 | 90 | ||
GenBank mRNA | AK343395.1 | - | 1 - 2 | 178 | 90 | |
AK392240.1 | - | 1 - 2 | 145 | 90 | ||
AK399833.1 | - | 1 - 2 | 195 | 90 | ||
AY610251.1 | - | 1 - 2 | 126 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
