Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244655.1 | NP_001231584.1 | 3 - 4 | 398 | 106 | |
GenBank mRNA | AK233902.1 | - | 3 - 4 | 398 | 106 | |
AY609543.1 | - | 3 - 4 | 383 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
