Assay Details
Gene Symbol: FLVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142840.1 | NP_001136312.1 | 3 - 4 | 1167 | 70 | |
GenBank mRNA | AK389329.1 | - | 3 - 4 | 626 | 70 | |
FJ236708.1 | - | 3 - 4 | 1167 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
