Assay Details
Gene Symbol: GZMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198926.1 | NP_001185855.1 | 3 - 4 | 361 | 80 | |
GenBank mRNA | AK346600.1 | - | 3 - 4 | 385 | 80 | |
HQ225831.1 | - | 3 - 4 | 361 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
