Assay Details
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204369.1 | NP_001191298.1 | 2 - 3 | 196 | 93 | |
XM_005655131.3 | XP_005655188.1 | 357 | 93 | |||
XM_005655133.3 | XP_005655190.1 | 146 | 93 | |||
XM_021085705.1 | XP_020941364.1 | 7453 | 93 | |||
XM_021085706.1 | XP_020941365.1 | 223 | 93 | |||
XM_021085707.1 | XP_020941366.1 | 7453 | 93 | |||
GenBank mRNA | AK231371.1 | - | 2 - 3 | 480 | 93 | |
AK235702.1 | - | 3 - 4 | 284 | 93 | ||
AK239959.1 | - | 2 - 3 | 202 | 93 | ||
AK347895.1 | - | 2 - 3 | 196 | 93 | ||
AK396107.1 | - | 2 - 3 | 194 | 93 | ||
AY610452.1 | - | 2 - 3 | 148 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
