Assay Details
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204369.1 | NP_001191298.1 | 4 - 5 | 350 | 81 | |
XM_005655131.3 | XP_005655188.1 | 511 | 81 | |||
XM_005655133.3 | XP_005655190.1 | 300 | 81 | |||
XM_021085704.1 | XP_020941363.1 | 544 | 81 | |||
XM_021085705.1 | XP_020941364.1 | 7607 | 81 | |||
XM_021085706.1 | XP_020941365.1 | 377 | 81 | |||
XM_021085707.1 | XP_020941366.1 | 7607 | 81 | |||
GenBank mRNA | AK231371.1 | - | 4 - 5 | 634 | 81 | |
AK235702.1 | - | 5 - 6 | 438 | 81 | ||
AK239959.1 | - | 4 - 5 | 356 | 81 | ||
AK347895.1 | - | 4 - 5 | 350 | 81 | ||
AK396107.1 | - | 4 - 5 | 348 | 81 | ||
AY610452.1 | - | 4 - 5 | 302 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
