Assay Details
Gene Symbol: BHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204771.1 | NP_001191700.1 | 4 - 5 | 463 | 79 | |
GenBank mRNA | AK232714.1 | - | 4 - 5 | 462 | 79 | |
HQ130334.1 | - | 4 - 5 | 463 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
