Assay Details
Gene Symbol: PLA2G2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244874.1 | NP_001231803.1 | 4 - 5 | 526 | 96 | |
GenBank mRNA | AK349876.1 | - | 4 - 5 | 528 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
