Assay Details
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244925.1 | NP_001231854.1 | 2 - 3 | 124 | 77 | |
XM_005655249.3 | XP_005655306.1 | 263 | 77 | |||
GenBank mRNA | AK346618.1 | - | 2 - 3 | 124 | 77 | |
AK392665.1 | - | 2 - 3 | 125 | 77 | ||
AK395675.1 | - | 2 - 3 | 143 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
