Assay Details
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244943.1 | NP_001231872.1 | 7 - 8 | 866 | 68 | |
XM_005665240.3 | XP_005665297.2 | 916 | 68 | |||
GenBank mRNA | AK349783.1 | - | 7 - 8 | 866 | 68 | |
AK393463.1 | - | 7 - 8 | 885 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
