Assay Details
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245976.1 | NP_001232905.1 | 2 - 3 | 209 | 104 | |
XM_021068454.1 | XP_020924113.1 | 273 | 104 | |||
XM_021068455.1 | XP_020924114.1 | 550 | 104 | |||
XM_021068456.1 | XP_020924115.1 | 343 | 104 | |||
XM_021068457.1 | XP_020924116.1 | 306 | 104 | |||
XM_021068458.1 | XP_020924117.1 | 646 | 104 | |||
XM_021068459.1 | XP_020924118.1 | 236 | 104 | |||
GenBank mRNA | AK393600.1 | - | 2 - 3 | 204 | 104 | |
JF271685.1 | - | 2 - 3 | 209 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
