Assay Details
Gene Symbol: APC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206430.1 | NP_001193359.1 | 13 - 14 | 1764 | 95 | |
GenBank mRNA | HQ288551.1 | - | 13 - 14 | 1764 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
