Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005663592.3 | XP_005663649.2 | 1124 | 61 | ||
XM_013997295.2 | XP_013852749.2 | 1136 | 61 | |||
XM_021090086.1 | XP_020945745.1 | 1124 | 61 | |||
XM_021090088.1 | XP_020945747.1 | 1124 | 61 | |||
XM_021090089.1 | XP_020945748.1 | 2441 | 61 | |||
GenBank mRNA | AK349445.1 | - | 7 - 8 | 1022 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
